missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL1RL1/ST2 Human ELISA Kit

Sandwich ELISA Kit

£584.00 - £3950.00

Specifications

Accession Number Q01638
Assay Sensitivity 30.5 pg/mL
Description Human Growth Stimulation-Expressed Protein ST2 ELISA Kit
Conjugate Biotin
Detection Method Fluorescence
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15904224
View Documents
Invitrogen™
EHIAST2
96 Tests
£584.00
Pack of 96
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
 
17195459
View Documents
Invitrogen™
EHIAST2X5
5 x 96 Tests
£2310.00
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
17895107
View Documents
Invitrogen™
EHIAST2X10
£3950.00
Pack of 10
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
Description

Description

The Human IL1RL1/ST2 ELISA quantitates Hu IL1RL1 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL1RL1. Principle of the method The Human IL1RL1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

ST2 is a member of the interleukin 1 receptor family. Studies of ST2 in mouse suggested that this receptor can be induced by proinflammatory stimuli, and may be involved in the function of helper T cells. ST2 belongs to the interleukin-1 receptor family with three Ig-like C2-type (immunoglobulin-like) domains and a TIR domain. ST2 is a receptor for interleukin-33 (IL-33) which upon stimulation recruits the signaling molecules MYD88, IRAK1, IRAK4, and TRAF6, followed by phosphorylation of MAPK3/ERK1 and/or MAPK1/ERK2, MAPK14, and MAPK8. Reports also suggest its possible role in helper T-cell function as well as in cell growth. Three isoforms of ST2 have been reported where isoform C is a membrane form and isoform B is a secreted form. ST2 may be induced by proinflammatory stimuli, and may also be found in serum of autoimmune disease. Different isoforms are restricted to different tissues while high expression of this protein is evident in kidney, lung, placenta, stomach, skeletal muscle, colon and small intestine.
Specifications

Specifications

Q01638
Human Growth Stimulation-Expressed Protein ST2 ELISA Kit
Fluorescence
Plasma, Serum, Supernatant
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
9173
8.5%
HRP
Inflammation
Human
3 hr.
30.5 pg/mL
Biotin
ELISA Kit
Human
Colorimetric Microplate Reader
IL1RL1
4.3%
RUO
-20°C
1 hr. 20 min.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.