missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Exonuclease I
Catalyzes sequential removal of nucleotides 3' end of single-stranded DNA molecule
Brand: Cytiva E70073Z
This item is not returnable.
View return policy
Description
- Monomeric and bacterial
- Molecular weight: 54.5kd
- Expressed in E.Coli
- Inactivated by heating at 80°C for 15 minutes
- Active in PCR buffers
- Activity: 10U/μL
- Free of contaminating endonucleases, double-stranded exonucleases and ribonucleases
- >95% pure by SDS-PAGE
Specifications
>95% by SDS-PAGE | |
2500 U | |
54.5kd |
Bacteria | |
Exonuclease I |