missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Exonuclease I
Catalyzes sequential removal of nucleotides 3' end of single-stranded DNA molecule
£344.00 - £584.00
Specifications
Purity | >95% by SDS-PAGE |
---|---|
Source | Bacteria |
Product Type | Exonuclease I |
Molecular Weight (g/mol) | 54.5kd |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10228854
|
Cytiva
E70073Z |
2500 U |
£344.00
2500 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10300175
|
Cytiva
E70073X |
5000 U |
£584.00
5000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Monomeric and bacterial
- Molecular weight: 54.5kd
- Expressed in E.Coli
- Inactivated by heating at 80°C for 15 minutes
- Active in PCR buffers
- Activity: 10U/μL
- Free of contaminating endonucleases, double-stranded exonucleases and ribonucleases
- >95% pure by SDS-PAGE
Specifications
>95% by SDS-PAGE | |
Bacteria | |
Exonuclease I | |
54.5kd |