Learn More
Cytiva ExoProStar™ 1-step
Use for enzymatic PCR clean-up. Cytiva Lifescience™ Illustra ExoProStar™ 1-step with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs.
Brand: Cytiva US77720
1170.40 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : GH
Additional Details : Weight : 0.02700kg
Description
- One tube with one mix of enzymes delivering PCR and sequence reaction clean-up
- Just one pipetting step to set up your reaction
- Fast 30-minute, hands-free protocol
- 100% recovery of PCR product
- Scalable for different reaction sizes
- Automation friendly
Specifications
Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs. For maximum convenience comes in a single tube. | |
Remove unincorporated primers and dNTPs | |
illustra ExoProStar 1-Step | |
30min. |
Enzymatic PCR Clean-up Technology | |
2000 | |
1 Each | |
100% recovery of PCR product |