Learn More
Cytiva GenomiPhi HY DNA Amplification Kit
Highly efficient and representative whole-genome amplification yielding 40 to 50μg from nanogram amounts of DNA sample
Marque: Cytiva 25-6600-25
Code : GE
Informations supplémentaires : CAS Number : 56-81-5 Weight : 0.20600kg
Includes: Sample Buffer, reaction buffer, enzyme mix, and control λ DNA
Description
- Starting material can be purified DNA or nonpurified cell lysates
- Simple protocol provides typical DNA of 40 to 50μg, average lengths >10kb
- Less hands-on time compared to traditional isolation methods
- Representative amplification of the whole genome
- Automation-friendly protocol with great reproducibility
- High-quality DNA for array CGH, high-throughput genotyping, DNA archival, PCR, restriction enzyme digestion, hybridization, and cloning
- Contains all of the components necessary for genomic DNA preparation by isothermal strand displacement amplification
Spécification
Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.Thaw components on ice and maintain at 0°C to 4°C during handling. | |
Genomic DNA Amplification | |
1000 | |
> 10 kb |
Illustra GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 50 μg yield from nanogram amounts of DNA sample. | |
10 x 2.25mL Sample Buffer, 10 x 2.25mL Reaction Buffer, 10 x 250μL Enzyme mix, 10 x 20μL Control DNA (lambda) | |
illustra GenomiPhi HY DNA Amplification Kit | |
4 h |