missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GenomiPhi HY DNA Amplification Kit
Highly efficient and representative whole-genome amplification yielding 40 to 50μg from nanogram amounts of DNA sample
£590.00 - £17300.00
Specifications
Content And Storage | Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.Thaw components on ice and maintain at 0°C to 4°C during handling. |
---|---|
Description | Illustra GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 50 μg yield from nanogram amounts of DNA sample. |
For Use With (Application) | Genomic DNA Amplification |
Product Type | illustra GenomiPhi HY DNA Amplification Kit |
Size | > 10 kb |
Product Code | Brand | Includes | No. of Reactions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | No. of Reactions | Price | Quantity & Availability | ||||
10247754
|
Cytiva
25-6600-20 |
2.25mL Sample Buffer, 2.25mL Reaction Buffer, 250μL Enzyme mix, 20mL Control DNA (lambda) | 100 |
£1830.00
100 reactions |
Estimated Shipment: 07-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10074774
|
Cytiva
25-6600-22 |
565μL Sample Buffer, 565μL Reaction Buffer, 65μL Enzyme mix, 20μL Control DNA (lambda) | 25 |
£590.00
25 reactions |
Estimated Shipment: 07-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10714357
|
Cytiva
25-6600-25 |
10 x 2.25mL Sample Buffer, 10 x 2.25mL Reaction Buffer, 10 x 250μL Enzyme mix, 10 x 20μL Control DNA (lambda) | 1000 |
£17300.00
1000 reactions |
Estimated Shipment: 07-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Starting material can be purified DNA or nonpurified cell lysates
- Simple protocol provides typical DNA of 40 to 50μg, average lengths >10kb
- Less hands-on time compared to traditional isolation methods
- Representative amplification of the whole genome
- Automation-friendly protocol with great reproducibility
- High-quality DNA for array CGH, high-throughput genotyping, DNA archival, PCR, restriction enzyme digestion, hybridization, and cloning
- Contains all of the components necessary for genomic DNA preparation by isothermal strand displacement amplification
Specifications
Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.Thaw components on ice and maintain at 0°C to 4°C during handling. | |
Genomic DNA Amplification | |
> 10 kb |
Illustra GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 50 μg yield from nanogram amounts of DNA sample. | |
illustra GenomiPhi HY DNA Amplification Kit | |
4 h |