missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Microspin G-50 Columns

Designed to allow the rapid purification of DNA for use in a wide range of applications using typical microfuge equipment. Cytiva illustra™ Microspin G-50 Columns will not remove or denature enzyme.

Brand:  Cytiva 27-5330-01

176.89 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : GH

Additional Details : CAS Number : 9048-46-8 Weight : 0.18720kg

 View more versions of this product

Product Code. 11753309

  • £199.00 / Pack of 50
Estimated Shipment: 02-08-2024
to see stock.

Description

Description

  • Optimal sample volumes in the range of 12-50μ
  • For rapid purification of labeled DNA (>20 bases) from smaller molecules in solution
  • Columns are prepacked with Sephadex G-50 Grade equilibrated in TE buffer with 0.05% Kathon CG/ICP Biocide
  • illustra AutoSeq™ and illustra ProbeQuant™ G-50 Micro Columns are specifically designed for dye terminator removal and labelled probe purification respectively and offer optimized performance in those applications
  • Can also be used for buffer exchange/desalting of enzymatic reactions (10 to 100 μL) following heat denaturation and phenol extraction
  • Product recovery will be at least 10% lower than that from ProbeQuant G-50 Micro Columns.
  • For rapid purification of labeled DNA (>20 bases) from unincorporated labeled nucleotides in a volume of 25 to 50 μl using spin-column chromatography

 

Desalting, buffer exchange; Removal of dye terminators from cycle sequencing reactions; Removal of labeled nucleotides from DNA labeling reactions

Specifications

Specifications

Desalting, Buffer exchange, removal of dye terminators from cycle sequencing reactions, removal of labeled, Nucleotides in 25 to 50μL from DNA labeling reactions
Sephadex™ G-50 DNA grade
50 Preps
Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers