missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Microspin G-50 Columns
Designed to allow the rapid purification of DNA for use in a wide range of applications using typical microfuge equipment. Cytiva illustra™ Microspin G-50 Columns will not remove or denature enzyme.
£199.00 - £804.00
Specifications
For Use With (Application) | Desalting, Buffer exchange, removal of dye terminators from cycle sequencing reactions, removal of labeled, Nucleotides in 25 to 50μL from DNA labeling reactions |
---|---|
Includes | Sephadex™ G-50 DNA grade |
Product Type | Column |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11753309
|
Cytiva
27-5330-01 |
50 Preps |
£199.00
Pack of 50 |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11763309
|
Cytiva
27-5330-02 |
250 Preps |
£804.00
Pack of 250 |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Optimal sample volumes in the range of 12-50μ
- For rapid purification of labeled DNA (>20 bases) from smaller molecules in solution
- Columns are prepacked with Sephadex G-50 Grade equilibrated in TE buffer with 0.05% Kathon CG/ICP Biocide
- illustra AutoSeq™ and illustra ProbeQuant™ G-50 Micro Columns are specifically designed for dye terminator removal and labelled probe purification respectively and offer optimized performance in those applications
- Can also be used for buffer exchange/desalting of enzymatic reactions (10 to 100 μL) following heat denaturation and phenol extraction
- Product recovery will be at least 10% lower than that from ProbeQuant G-50 Micro Columns.
- For rapid purification of labeled DNA (>20 bases) from unincorporated labeled nucleotides in a volume of 25 to 50 μl using spin-column chromatography
Desalting, buffer exchange; Removal of dye terminators from cycle sequencing reactions; Removal of labeled nucleotides from DNA labeling reactions
Order Info
Conveniently prepacked with Sephadex G-50 DNA Grade pre-equilibrated in TE buffer with 0.05% Kathon™ CG/ICP Biocide and collection tubes
Specifications
Desalting, Buffer exchange, removal of dye terminators from cycle sequencing reactions, removal of labeled, Nucleotides in 25 to 50μL from DNA labeling reactions | |
Sephadex™ G-50 DNA grade | |
Column |