missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Shrimp Alkaline Phosphatase

Catalyzes removal of 5'- and 3'-phosphate groups from DNA, RNA, nucleotides, and proteins

Brand:  Cytiva E70092X

 View more versions of this product

Product Code. 10626895

  • £630.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 10x reaction buffer: 200mM Tris-HCl, pH 8.0, 100mM MgCl2; Dilution buffer: 50mM Tris-HCl, pH 8.0
This item is not returnable. View return policy

Description

Description

  • Bacterial
  • Expressed in E.Coli
  • Inactivated by heating at 75°C for 5 minutes
  • Active in PCR buffers and restriction endonuclease buffers
  • Activity: 1U/μL
  • Purified to homogeneity
  • Free of all contaminating endonucleases, exonucleases and ribonucleases
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Purified to homogeneity
Bacteria
5000 U
Alkaline Phosphatase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers