missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Shrimp Alkaline Phosphatase
Catalyzes removal of 5'- and 3'-phosphate groups from DNA, RNA, nucleotides, and proteins
£114.42 - £618.00
Specifications
Purity | Purified to homogeneity |
---|---|
Source | Bacteria |
Product Type | Alkaline Phosphatase |
Includes: 10x reaction buffer: 200mM Tris-HCl, pH 8.0, 100mM MgCl2; Dilution buffer: 50mM Tris-HCl, pH 8.0
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10723597
|
Cytiva
E70092Y |
500 U |
£114.42
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10723787
|
Cytiva
E70092Z |
1000 U |
£184.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10626895
|
Cytiva
E70092X |
5000 U |
£618.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Bacterial
- Expressed in E.Coli
- Inactivated by heating at 75°C for 5 minutes
- Active in PCR buffers and restriction endonuclease buffers
- Activity: 1U/μL
- Purified to homogeneity
- Free of all contaminating endonucleases, exonucleases and ribonucleases
Specifications
Purified to homogeneity | |
Bacteria | |
Alkaline Phosphatase |