Learn More
Cytiva TempliPhi™ 100/500 Amplification Kit
Use amplified DNA directly for cycle sequencing without purification. Cytiva illustra TempliPhi DNA Amplification Kits efficiently prepare DNA sequencing templates from circular DNA.
Brand: Cytiva 25-6400-50
1043.44 GBP valid until 2024-12-31
Use promo code "21949" to get your promotional price.
Includes: Sample buffer, Reaction buffer, Enzyme mix, and Positive control pUC19 DNA (2ng/μL)
Description
- Generate microgram quantities of template DNA from picogram amounts of starting material
- Completed reactions contain uniform quantities of DNA irrespective of input amount, allowing for use directly in sequencing reactions without quantification
- Amplify DNA from bacterial or M13 liquid cultures, colonies, plaques, glycerol stocks, or purified circular (plasmid or M13) DNA without the need for purification
- Simple protocol reduces time, labor, and consumables needed for template preparation, and workflow enables easy automation
- Contain separate, unmixed components for enhanced stability
Specifications
The product can be stored for 2 months or less at -20°C. For storage longer than 2 months, store at -70°CThaw components on ice and maintain at 0–4°C during handling. DO NOT warm above 4°C prior to amplification | |
Sequencing Template Amplification Kits and Reagents | |
500 | |
4-18 h depending on starting material |
illustra TempliPhi DNA Amplification Kits efficiently prepare DNA sequencing templates from circular DNA. | |
0.5mL Sample Buffer, 0.5mL Reaction Buffer, 20μL Enzyme mix and 50μL positive control pUC19 DNA | |
illustra TempliPhi 500 Amplification Kit |