missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva TempliPhi™ 100/500 Amplification Kit
Use amplified DNA directly for cycle sequencing without purification. Cytiva illustra TempliPhi DNA Amplification Kits efficiently prepare DNA sequencing templates from circular DNA.
£237.00 - £1141.00
Specifications
Content And Storage | The product can be stored for 2 months or less at -20°C. For storage longer than 2 months, store at -70°CThaw components on ice and maintain at 0–4°C during handling. DO NOT warm above 4°C prior to amplification |
---|---|
Description | illustra TempliPhi DNA Amplification Kits efficiently prepare DNA sequencing templates from circular DNA. |
For Use With (Application) | Sequencing Template Amplification Kits and Reagents |
Includes | 0.5mL Sample Buffer, 0.5mL Reaction Buffer, 20μL Enzyme mix and 50μL positive control pUC19 DNA |
Test Time | 4-18 h depending on starting material |
Includes: Sample buffer, Reaction buffer, Enzyme mix, and Positive control pUC19 DNA (2ng/μL)
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10794562
|
Cytiva
25640010 |
100 |
£237.00
1 set |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10281390
|
Cytiva
25-6400-50 |
500 |
£1141.00
Each |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Generate microgram quantities of template DNA from picogram amounts of starting material
- Completed reactions contain uniform quantities of DNA irrespective of input amount, allowing for use directly in sequencing reactions without quantification
- Amplify DNA from bacterial or M13 liquid cultures, colonies, plaques, glycerol stocks, or purified circular (plasmid or M13) DNA without the need for purification
- Simple protocol reduces time, labor, and consumables needed for template preparation, and workflow enables easy automation
- Contain separate, unmixed components for enhanced stability
Specifications
The product can be stored for 2 months or less at -20°C. For storage longer than 2 months, store at -70°CThaw components on ice and maintain at 0–4°C during handling. DO NOT warm above 4°C prior to amplification | |
Sequencing Template Amplification Kits and Reagents | |
4-18 h depending on starting material |
illustra TempliPhi DNA Amplification Kits efficiently prepare DNA sequencing templates from circular DNA. | |
0.5mL Sample Buffer, 0.5mL Reaction Buffer, 20μL Enzyme mix and 50μL positive control pUC19 DNA |