missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-iT™ Plant Tissue Hard-Set Mountant

Image-iT Plant Tissue Hard-Set Mountant is used for imaging, storage, and archiving of plant tissues optically cleared with Image-iT Plant Tissue Clearing Reagent.

Brand:  Invitrogen™ V11331

104.88 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 16329583

  • £138.00 / 30mL
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

These reagents are compatible with both fluorescence-based and colorimetric-based applications. Image-iT Plant Tissue mountant is based on Visikol™ Mount, which has a history of use in fluorescent and colorimetric studies with plants.

• Clears and mounts plant tissue for whole specimen imaging
• Hard-set mountant with refractive index of 1.415

TRUSTED_SUSTAINABILITY
Specifications

Specifications

30 mL
Image-iT™
30 mL
Room Temperature
Store at 15°C to 25°C. Do not freeze.
When stored as directed, product is stable for (at least 6 months after receipt.

)
Mountant
Plant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.