Learn More
Imidazole-2-carboxaldehyde, 97%, Thermo Scientific Chemicals
£61.60 - £237.00
Chemical Identifiers
CAS | 10111-08-7 |
---|---|
Molecular Formula | C4H4N2O |
Molecular Weight (g/mol) | 96.09 |
MDL Number | MFCD00003544 |
InChI Key | XYHKNCXZYYTLRG-UHFFFAOYSA-N |
Synonym | imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole |
PubChem CID | 24955 |
IUPAC Name | 1H-imidazole-2-carbaldehyde |
SMILES | O=CC1=NC=CN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11343297
|
Thermo Scientific Alfa Aesar
L13883.06 |
5 g |
£61.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11353297
|
Thermo Scientific Alfa Aesar
L13883.14 |
25 g |
£237.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Imidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsImidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.
Solubility
Soluble in water.
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
10111-08-7 | |
96.09 | |
XYHKNCXZYYTLRG-UHFFFAOYSA-N | |
24955 | |
O=CC1=NC=CN1 |
C4H4N2O | |
MFCD00003544 | |
imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole | |
1H-imidazole-2-carbaldehyde |
Specifications
10111-08-7 | |
C4H4N2O | |
4371302 | |
imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole | |
XYHKNCXZYYTLRG-UHFFFAOYSA-N | |
1H-imidazole-2-carbaldehyde | |
24955 | |
97% |
∽205°C (decomposition) | |
MFCD00003544 | |
Air sensitive | |
Soluble in water. | |
O=CC1=NC=CN1 | |
96.09 | |
96.09 | |
Imidazole-2-carboxaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only