missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ Maleimide-Activated BSA

Prepare hapten-carrier conjugates with cysteine-containing peptides and other thiolated antigens using this sulfhydryl-reactive bovine serum albumin.

Brand:  Thermo Scientific™ 77115

 View more versions of this product

Product Code. 10648304

  • £489.00 / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Upon receipt store product at 4°C. Product is shipped at ambient temperature.
10 mg
Imject™
Imject Maleimide Activated Bovine Serum Albumin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberYI385978 Certificate TypeCertificate of Analysis Date29/09/2023 Catalog Number77115
Lot NumberXL370647 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number77115
Lot NumberXF357241 Certificate TypeCertificate of Analysis Date14/07/2022 Catalog Number77115
Lot NumberWJ328762 Certificate TypeCertificate of Analysis Date06/11/2021 Catalog Number77115
Lot NumberVL319652 Certificate TypeCertificate of Analysis Date19/12/2020 Catalog Number77115
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Imject™ Maleimide-Activated BSA > Activated BSA; 10mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.