Learn More
Ebro Electronic™ Thermocouple Rod Probes
Rod probes for temperature measurement of air, ovens, fluid and gases.
Brand:  Ebro Electronic™ 1341-0607
 View more versions of this product
Product Code. 10464473
  • £111.00 / Each
Explore more special offers
Description
Description
For various applications in food, laboratory, industial use.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RoHS Directive 2011/65/EU
Single-use
15 mm
220 mm
Metal
TPN 140
0.24 kg
1100°C
-40 °F to +2012 °F
TFN 520
15 mm
3 mm
Measurement of air, oven, fluids and gas temperatures
Total or Partial
185 mm
4 sec.
NiCrNi rod
Immersion Probe
-40°C
-40°C to +1100°C
DIN IEC 584, Class 2