missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Immu-Mount™

Non-permanent mountant for use with frozen sectioning, immunoperoxidase techniques and immunofluorescent applications.

Brand:  Epredia™ 9990412

77.50 GBP valid until 2024-11-29
Use promo code "21706" to get your promotional price.



 View more versions of this product

Product Code. 10622689

  • £193.00 / Pack of 6
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Economical, organic, overnight-drying liquid coverslip
  • Aqueous base for immunostaining and other techniques
  • Non-fluorescing medium with a low viscosity
  • Resists fading and polymerizes quickly for easier handling of finished slides
  • Refractive index: 1.495 ±0.002 (liquid); 1.586 ± 0.002 (dry)
  • Viscosity: Low (120-160 cps at 24°C)
  • pH: 8.0 from 8.8 at 21 through 24°C
Specifications

Specifications

For use with frozen section, immunoperoxidase techniques, and immunofluorescent preparations
6 x 20 mL
Shandon Immu-mount
20 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : toluene, butyl methacylate, 2,6-di-tert-butyl-p-cresol, butyl benzyl phthalate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers