Learn More
Thermo Scientific™ Immuno Breakable Modules Clear
Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.
Brand: Thermo Scientific™ 95029390
329.94 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 04
Additional Details : Weight : 3.09000kg
Description
Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.
Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.
Features:
- For solid-phase immunoassays, e.g. ELISA
- Choice of four surfaces for optimal binding
- Free-standing modules are easy to handle
- Alphanumeric system for easy orientation
- Fits standard equipment
- High optical quality for reliable results
- Polystyrene
Choice of Surface:
- PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
- MediSorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
- MaxiSorp™, Immulon 4 HBX, and Enhanced Binding (EB) are hydrophilic and ideal for antibody sandwich assays
- MultiSorp™ are very hydrophilic and work well for glycans and water-soluble proteins
Specifications
Breakable Strips | |
No | |
High optical quality for reliable results | |
25 Pk., 50 Cases | |
Clear | |
Polystyrene | |
96 | |
Universal Binding | |
For adsorption of hydrophobic molecules | |
50 | |
Lipids |
Hydrophobic | |
Strip Plate | |
Colorimetric | |
Adsorption | |
Microplate Strip | |
400 μL | |
350 μL | |
Round, Flat Bottom (F-well) | |
50 | |
None |