missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Immuno Breakable Modules in White and Black
Keep wells in place with these breakable modules in white and black, ensuring fluorescence and luminescence assays are easy to handle.
Brand: Thermo Scientific™ 95029800
This item is not returnable.
View return policy
Description
Keep wells securely in place with Thermo Scientific™ Breakable Modules in White and Black, ensuring fluorescence and luminescence assays are easy to handle.
Features:- High-quality polystyrene wells
- Choice of two passive surfaces to enable assay optimization
- C8 Strips provide flexiblity with sample size and optimal washing performance
- Easy to handle; single well held as easily as strips
- Compatible with common instrumentation
Choice of Surface:
- Thermo Scientific™ PolySorp™ and Universal Binding (UB) are for adsorption of hydrophobic molecules.
- Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assays.
White:
- Opaque and reflective surface
- Low cross talk and minimal background
Black:
- Opaque and light-absorbing surface
- Minimum cross talk and back-scattered light
Note:
Availability may vary by country.
Specifications
Hydrophobic | |
Strip Plate | |
8 x 12 | |
Black | |
Polystyrene | |
350 μL | |
Flat Bottom (F-well) | |
50 | |
None |
No | |
Fluorescence | |
50 Boxes | |
Microplate Strip | |
96 | |
Universal Binding | |
Fluorescence | |
50 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction