missing translation for 'onlineSavingsMsg'
Learn More

Indium foil, 0.5mm (0.02in) thick, Puratronic™, 99.9975% (metals basis), Thermo Scientific Chemicals

£78.99 - £1006.01

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronicShow MoreShow Less
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11361179
SDS View Documents
Thermo Scientific Alfa Aesar
011384.FF
25 x 25 mm
£78.99
25mm
Please to purchase this item. Need a web account? Register with us today!
11345849
SDS View Documents
Thermo Scientific Alfa Aesar
011384.FI
50 x 50 mm
£170.85
Each
Please to purchase this item. Need a web account? Register with us today!
11351179
SDS View Documents
Thermo Scientific Alfa Aesar
011384.FY
50 x 100 mm
£342.71
Each
Please to purchase this item. Need a web account? Register with us today!
11341179
SDS View Documents
Thermo Scientific Alfa Aesar
011384.GK
100 x 200 mm
£1006.01
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-180-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium foil, 0.5mm (0.02in) thick, Puratronic™, 99.9975% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.