missing translation for 'onlineSavingsMsg'
Learn More
Indium(III) oxide, 99.994% (metals basis), Thermo Scientific Chemicals Product Code.: 11341918

Indium(III) oxide, 99.994% (metals basis), Thermo Scientific Chemicals

Product Code. 11341918
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11341918

Brand: Thermo Scientific Alfa Aesar 011853.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indium(III) oxide is used as a chemical reagent and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]

Specifications

Melting Point 850° volat.
Color Yellow
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.994% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 277.64
Odor Odorless
Density 7.18
Chemical Name or Material Indium(III) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT22K012 Certificate TypeCertificate of Analysis Date06/08/2024 Catalog Number011853.09,011853.30,011853.18
Lot NumberZ16I016 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number011853.09,011853.30,011853.18
Lot NumberZ20H047 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011853.09,011853.30,011853.18
Lot NumberH18K46 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011853.09,011853.30,011853.18
Lot NumberI09M18 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011853.09,011853.30,011853.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium(III) oxide, 99.994% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.