missing translation for 'onlineSavingsMsg'
Learn More
Indium shot, tear drop, 4mm (0.2in), 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11321648

Indium shot, tear drop, 4mm (0.2in), 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11321648
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11321648

Brand: Thermo Scientific Alfa Aesar 011027.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronicShow MoreShow Less
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Color Silver-White
Appearance Silvery-white shot
Density 7.31g/cm3 at 20°C
Assay Percent Range 99.99% (Metals basis)
Quantity 250 g
Odor Odorless
Form Shot
Chemical Name or Material Indium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY27J007 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number011027.09,011027.18,011027.30
Lot NumberP04I138 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011027.09,011027.18,011027.30,011029.03
Lot NumberY08H007 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011027.09,011027.18,011027.30,011029.03
Lot NumberQ29A010 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011027.09,011027.18,011027.30,011029.03
Lot NumberW28H014 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011027.09,011027.18,011027.30,011029.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium shot, tear drop, 4mm (0.2in), 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.