missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Water (I-Z)

European Standard Pipemarkers reinforce safety and aid maintenance.

Brand:  Brady™ N009387

Code : NEW

Additional Details : Weight : 0.10600kg

 View more versions of this product

Product Code. 15359465

  • £102.00 / 1 roll
Estimated Shipment: 15-08-2024
to see stock.
Add to basket

Description

Description

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications

Specifications

ROHS Environmental Compliance, EU Directive 2002/95/EC
Permanent Acrylic, Permanent, Pressure-sensitive
White on Green
Polyester
Treated water
English
30 Pk.
-40°C to +140°C
0.119 mm
33 m x 250 mm x 26 mm
33 m
250 mm
26 mm
Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Water - Treated water
Silicone
Roll
Pipe Marker
Pipe and valve marking
30 Markers/Roll
-40°F to +284°F
0.0047 in.
33 m x 250 mm
36.09 yd.
9.84 in.
1.02 in.
13.3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers