Learn More
Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Water (I-Z)
European Standard Pipemarkers reinforce safety and aid maintenance.
£102.00 - £154.00
Specifications
Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC |
---|---|
Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
Coating | Silicone |
Color | White on Green |
Format | Roll |
Product Code | Brand | Display Text | Height (Metric) | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Text | Height (Metric) | Width (Metric) | Price | Quantity & Availability | |||
15339425
|
Brady™
N009225 |
Industrial water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15349425
|
Brady™
N009226 |
Industrial water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15359425
|
Brady™
N009227 |
Industrial water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15349435
|
Brady™
N009272 |
Non-drinking water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329435
|
Brady™
N009270 |
Non-drinking water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15379445
|
Brady™
N009315 |
Process water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15339435
|
Brady™
N009271 |
Non-drinking water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15389445
|
Brady™
N009316 |
Process water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15399445
|
Brady™
N009317 |
Process water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15379315
|
Brady™
N008730 |
Raw water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15389315
|
Brady™
N008731 |
Raw water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15399315
|
Brady™
N008732 |
Raw water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15309485
|
Brady™
N009423 |
Return | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15319485
|
Brady™
N009424 |
Return | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329485
|
Brady™
N009425 |
Return | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15339395
|
Brady™
N009118 |
Soft water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329395
|
Brady™
N009117 |
Soft water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15349395
|
Brady™
N009119 |
Soft Water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15309275
|
Brady™
N008595 |
Supply | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329275
|
Brady™
N008597 |
Supply | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15319275
|
Brady™
N008596 |
Supply | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15359465
|
Brady™
N009387 |
Treated water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15369465
|
Brady™
N009388 |
Treated water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15379465
|
Brady™
N009389 |
Treated water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15319475
|
Brady™
N009396 |
Waste water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15329475
|
Brady™
N009397 |
Waste water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15339475
|
Brady™
N009398 |
Waste water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15349305
|
Brady™
N008685 |
Water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15359305
|
Brady™
N008686 |
Water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15369305
|
Brady™
N008687 |
Water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15379355
|
Brady™
N009000 |
Well water | 26 mm | 250 mm |
£102.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15389355
|
Brady™
N009001 |
Well water | 37 mm | 355 mm |
£122.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
15399355
|
Brady™
N009002 |
Well water | 52 mm | 450 mm |
£154.00
1 roll |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC | |
Silicone | |
Roll | |
Pipe Marker | |
English | |
30 Pk. | |
-40°C to +140°C | |
0.119 mm | |
33 m |
Permanent Acrylic, Permanent, Pressure-sensitive | |
White on Green | |
Polyester | |
Pipe and valve marking | |
30 Markers/Roll | |
-40°F to +284°F | |
0.0047 in. | |
36.09 yd. |