missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Water (A-H)

European Standard Pipemarkers reinforce safety and aid maintenance.

Brand:  Brady™ N009161

Code : NEW

Additional Details : Weight : 0.01500kg

 View more versions of this product

Product Code. 15399405

  • £12.55 / Pack of 2
Estimated Shipment: 15-08-2024
to see stock.
Add to basket

Description

Description

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications

Specifications

ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Permanent Acrylic, Permanent, Pressure-sensitive
Gloss
Polyester
Cold water supply
2
2 Markers/Card
-40°F to +230°F
0.0039 in.
52 x 450 mm
52 mm
450 mm
Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Water - Cold water supply
White on Green
Card
Pipe Marker
Pipe and valve marking
English
2 Pk.
-40°C to +110°C
0.099 mm
2.05 in.
17.72 in.
22.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers