Learn More
Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Water (A-H)
European Standard Pipemarkers reinforce safety and aid maintenance.
£12.30 - £15.25
Specifications
Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 |
---|---|
Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
Color | White on Green |
Finish | Gloss |
Format | Card |
Product Code | Brand | Width (Metric) | Display Text | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Display Text | Price | Quantity & Availability | ||||
15379305
|
Brady™
N008691 |
250 mm | Boiler feed water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15399305
|
Brady™
N008693 |
450 mm | Boiler feed water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15399455
|
Brady™
N009357 |
250 mm | Brine water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15319465
|
Brady™
N009359 |
450 mm | Brine water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15319405
|
Brady™
N009141 |
250 mm | Canal water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15339405
|
Brady™
N009143 |
450 mm | Canal water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15339455
|
Brady™
N009339 |
250 mm | Chilled water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15359455
|
Brady™
N009341 |
450 mm | Chilled water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15359335
|
Brady™
N008862 |
250 mm | Chlorated water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15319375
|
Brady™
N009044 |
450 mm | City water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15399365
|
Brady™
N009042 |
250 mm | City water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15379335
|
Brady™
N008864 |
450 mm | Chlorated water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15339415
|
Brady™
N009177 |
250 mm | Cold water return |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15359415
|
Brady™
N009179 |
450 mm | Cold water return |
£12.55
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15399405
|
Brady™
N009161 |
450 mm | Cold water supply |
£12.55
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15379405
|
Brady™
N009159 |
250 mm | Cold water supply |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15329295
|
Brady™
N008655 |
250 mm | Condensate |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15349295
|
Brady™
N008657 |
450 mm | Condensate |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15319345
|
Brady™
N008880 |
250 mm | Condenser water return |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15339345
|
Brady™
N008882 |
450 mm | Condenser water return |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15389345
|
Brady™
N008898 |
250 mm | Condenser water supply |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15309355
|
Brady™
N008900 |
450 mm | Condenser water supply |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15349345
|
Brady™
N008889 |
250 mm | Condenser water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15379345
|
Brady™
N008891 |
450 mm | Condenser water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15309365
|
Brady™
N009006 |
250 mm | Cooling water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15329365
|
Brady™
N009008 |
450 mm | Cooling water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15329375
|
Brady™
N009060 |
250 mm | Decarbonatized Water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15349375
|
Brady™
N009062 |
450 mm | Decarbonatized Water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15359375
|
Brady™
N009069 |
250 mm | Deionized water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15389375
|
Brady™
N009078 |
250 mm | Demineralised Water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15379375
|
Brady™
N009071 |
450 mm | Deionized water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15319385
|
Brady™
N009080 |
450 mm | Demineralised Water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15359385
|
Brady™
N009096 |
250 mm | Distilled water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15379385
|
Brady™
N009098 |
450 mm | Distilled water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15389435
|
Brady™
N009303 |
250 mm | Drinking water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15309445
|
Brady™
N009305 |
450 mm | Drinking water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15309405
|
Brady™
N009134 |
450 mm | Filtered water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15389395
|
Brady™
N009132 |
250 mm | Filtered water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15329285
|
Brady™
N008619 |
250 mm | Heating return |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15369275
|
Brady™
N008610 |
250 mm | Heating supply |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15349285
|
Brady™
N008621 |
450 mm | Heating return |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15319355
|
Brady™
N008916 |
250 mm | Heating water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15389275
|
Brady™
N008612 |
450 mm | Heating supply |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15339355
|
Brady™
N008918 |
450 mm | Heating water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15339275
|
Brady™
N008601 |
250 mm | Heating |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15319335
|
Brady™
N008837 |
450 mm | Hot water return |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15359275
|
Brady™
N008603 |
450 mm | Heating |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15399325
|
Brady™
N008835 |
250 mm | Hot water return |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15309325
|
Brady™
N008736 |
250 mm | Hot water |
£15.25
Pack of 4 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
15329325
|
Brady™
N008738 |
450 mm | Hot water |
£12.30
Pack of 2 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 | |
White on Green | |
Card | |
Pipe Marker | |
English | |
-40°C to +110°C | |
0.099 mm |
Permanent Acrylic, Permanent, Pressure-sensitive | |
Gloss | |
Polyester | |
Pipe and valve marking | |
-40°F to +230°F | |
0.0039 in. |