Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F1
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
Brand: Fisherbrand™ FB67776
Description
- Class F1 stainless steel calibration weight
- In compliance with OIML R111
Specifications
F1 | |
1 kg | |
Cylindrical |
Stainless Steel | |
1 kg |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F1 > 1kg
Spot an opportunity for improvement?Share a Content Correction