missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Teknon™ Biocleanse Disinfectant
Ready to use deodorizing cleaner,degreaser, disinfectant and fungicide for use in scientific, medical and biotechnology environments to combat biological hazards. Teknon™ Biocleanse Disinfectant is effective against a wide range of micro-organisms.
Brand: Teknon TK210/T
This item is not returnable.
View return policy
Description
- Effective against a wide range of micro-organisms including Pseudomonas aeruginosa, Staphylococcus aureus, Enterococcus hirae, E.coli, MRSA, VRE, Candida albicans, Penicillium spp, HIV, HBV, HCV, Swine flu (H1N1), Avian flu (diseases of poultry), Norovirus, Clostridium difficile, Mycobacterium tuberculosis and other mycobacteria. Full efficacy data is available on request.
- Use as supplied. Apply by spray, mop, soaking or using ultrasonic agitation. Rinse cleaned surfaces with water if necessary. Minimise contact with sensitive surfaces such as aluminum and polycarbonate.
- Safe to use on ferrous and non-ferrous metals, ceramics, glassware and plastics.
- The product is equivalent to a 5% solution of Biocleanse concentrate.
- Free from sodium hypochlorite, phosphates and enzymes.
- Effectively removes blood, fat, proteins, grease and other organic and non-organic contaminants while simultaneously disinfecting the treated surface.
- Meets the requirements of BS EN 1276 and is effective against MRSA, HIV, HBV, Avian Flu,E. coli, Salmonella, Listeria, Candida, Penicillium, and C. diff.
- Under COSHH Regulations, the product does not require a maximum exposure limit and is not irritating to skin.
- Ready to use, non tainting and ready to use deodorizing cleaner, degreaser, disinfectant and fungicide for use in scientific, medical and biotechnology environments to combat biological hazards.
- Store in a cool, dry place protected from frost and away from acids and strong oxidizing agents.
- Store upright in original containers
- Shelf life: 24 months in good conditions
Specifications
Bioicidal Agent | |
Light Blue | |
Alkaline sequestrant (0-1%), nonionic surfactant (0-1%), benzalkonium chloride (0-1%) | |
1.001 at 20°C | |
500 mL |
7 (1% aq. soln.) | |
100°C | |
Store in a cool, dry place protected from frost and away from acids and strong oxidizing agents. Store upright in original containers. | |
For use in scientific, medical and biotechnology environments to combat biological hazards |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Teknon™ Biocleanse Disinfectant > Quantity: 500mL
Spot an opportunity for improvement?Share a Content Correction