Learn More
Fisherbrand™ Aluminum Foil, Standard-Gauge Roll
For sealing, weighing, and transferring samples
Brand:  Fisherbrand™ 01213101
 View more versions of this product
Product Code. 11749408
  • £15.91 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Ideal for weighing, sealing, and tranferring your samples.
  • Packed in individual dispenser box with cutter bar
  • Heavy-gauge rolls offer greater folding strength, good for covering sharp edges
Specifications
Specifications
Sealing, Weighing, Transferring Samples
Foil
30.5cm
Aluminum
197 ft.
12 in.
60 m
18 μm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Aluminum Foil, Standard-Gauge Roll > Standard-gauge roll; Width: 12 in. (30.4cm); Length: 200 ft. (60.96m)