Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F1 with certificate
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
Brand:  Fisherbrand™ FB67906
 Afficher plus de versions de ce produit
Code produit. 15555189
  • £1075.00 / 1 unit
Estimated Shipment: 09-01-2025
to see stock.
Découvrez plus d'offres spéciales
Ajouter au panier
Description
Description
  • Class F1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
Spécification
Spécification
F1
10 kg
Cylindrical
Stainless Steel
10 kg
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats