missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Individually Wrapped Sterile Dippers

Ideal for safe sample collection and transportation in a single container, eliminates the risk of cross-contamination.

Brand:  Wheaton™ 327135

Additional Details : Weight : 2.25000kg

 View more versions of this product

Product Code. 16671784

  • £153.00 / Pack of 50
Estimated Shipment: 28-11-2024
to see stock.
Add to basket

Description

Description

  • Available in 3 sizes: 30 mL, 100 mL, and 250 mL
  • Supports food industry and pharmaceutical requirements
  • Tested in accordance with BS EN 14254 for safe, leak-free performance
  • Handle neatly snaps off after sample collection for ease of transportation
  • Long handle length accommodates a majority of sampling applications
  • Choice of clear and blue polystyrene
  • Sterilized by gamma irradiation
  • Individually wrapped
Specifications

Specifications

Dipper
Metal
Blue
385 mm
Individually Wrapped
100 mL
50/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers