missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Injection Port Liners with CarboFrit for Thermo Scientific™ Trace and Focus SSL Instruments

Ensure a high degree of reproducibility with Thermo Scientific Injection Port Liners with CarboFrit for Thermo Scientific Trace and Focus SSL Instruments.

Brand:  Thermo Scientific™ 453T2131

 View more versions of this product

Product Code. 10197594

  • £351.75 / Pack of 5
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Ensure a high degree of reproducibility with Thermo Scientific Injection Port Liners with CarboFrit for Thermo Scientific Trace and Focus SSL Instruments. Highly deactivated and produced to exacting tolerances.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

453T2131
105 mm
Split Straight Liner
8 mm
5
CarboFrit
5 mm
Split Straight Liner
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers