missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Expert pH Electrodes

A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.

Brand:  METTLER TOLEDO™ 51344104

378.75 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 10594925

  • £576.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Ideal for critical samples such as suspensions or samples of unknown composition. Mettler Toledo Expert Series pH and pH/ATC Combination Electrodes with open junction puts the reference electrode in direct contact with the measured medium, eliminating the possibility of contamination or blockage.

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Features a PEEK plastic shaft for harsh environments and requires no maintenance
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • General Lab
  • Beverages
  • Waste water
  • Suspended solids
  • Environmental
  • TRIS compatible

Accessories & Care

  • Open Junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BNC/RCA IP67
PEEK
0°C to 100°C
Xerolyt Polymer
Fixed Cable to sensor
U Glass
NTC 30 kilohm
0°C to 100°C
32.8 ft.
10 m
12 mm
Open
0 to 14
Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology
In harsh environments, such as in near-processor outdoor areas
ARGENTHAL
InLab™ Expert Go-ISM (10 m)
32°F to 212°F
4.72 in.
120 mm
0.47 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
METTLER TOLEDO™ InLab™ Expert pH Electrodes > Cable length: 10m

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.