Learn More
METTLER TOLEDO™ InLab™ Power pH Electrodes
A glass pH electrode with an integrated temperature sensor featuring the SteadyForce™ reference system, which is pressurized to prevent the junction from clogging.
Brand: METTLER TOLEDO™ 51344211
613.31 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : 59
Additional Details : Weight : 0.13000kg
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Corrosive acids and bases
- Protein-rich samples
- Dairy
- High pH values
- High temperature
- Junction-clogging samples
- Polymeric dispersions
- TRIS compatible
Accessories & Care
- Non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
pH Electrode InLab Power Pro-ISM | |
Ceramic | |
0 to 12 | |
Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | |
Multi-Pin to BNC/RCA (30281896) | |
A41 Glass | |
NTC 30 kω | |
32°F to 266°F | |
6.7 in. | |
0.47 in. |
Multi-Pin | |
Glass | |
0°C to 130°C | |
DPA Gel | |
SteadyForce | |
InLab™ Power Pro-ISM | |
3 mol/L KCl | |
0°C to 130°C | |
170 mm | |
12 mm |