missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Power pH Electrodes
A glass pH electrode with an integrated temperature sensor featuring the SteadyForce™ reference system, which is pressurized to prevent the junction from clogging.
£455.00 - £725.00
Specifications
Junction | Ceramic |
---|---|
Material | Glass |
pH Range | 0 to 12 |
Temperature (Metric) Operating | 0°C to 130°C |
Filling Solution Type | DPA Gel |
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
10192003
|
METTLER TOLEDO™
51343110 |
InLab™ Power | Combination Electrode | S7 | S7 to BNC (30281915) |
£455.00
Each |
In Stock
Log in to see stock. |
||
11304223
|
METTLER TOLEDO™
51344211 |
InLab™ Power Pro-ISM | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | Multi-Pin | Multi-Pin to BNC/RCA (30281896) |
£725.00
Each |
In Stock
Log in to see stock. |
||
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Corrosive acids and bases
- Protein-rich samples
- Dairy
- High pH values
- High temperature
- Junction-clogging samples
- Polymeric dispersions
- TRIS compatible
Accessories & Care
- Non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Ceramic | |
0 to 12 | |
DPA Gel | |
A41 Glass | |
32°F to 266°F | |
12 mm |
Glass | |
0°C to 130°C | |
SteadyForce | |
3 mol/L KCl | |
0.47 in. |