missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Solids pH Electrodes

A low maintenance glass pH electrode with an integrated temperature probe featuring a clog-free, open junction that eliminates the risk of protein fouling. Its spear shaped tip is designed for piercing solid and semi-solid food samples (IP67 rated).

Brand:  METTLER TOLEDO™ 51343156

367.07 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 11997771

  • £515.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Meats
  • Cheese
  • Seafood
  • Dough
  • Milk and cream
  • Butter, yogurt, ice cream
  • Fruits and vegetables
  • Ready-made meals
  • Solid and semi-solid food samples

Accessories & Care

  • Open junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

pH Electrode InLab Solids Go-ISM
Open
1 to 11
Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology
ARGENTHAL
InLab™ Solids Go-ISM
BNC IP67
Glass
0°C to 80°C
Fixed Cable to sensor
LoT Glass
1.80 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Up to 60% off
    Promo Code24119

    Expires2025-04-30

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    23

    Hours

    3

    Minutes

    33

    Seconds

    59
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
METTLER TOLEDO™ InLab™ Solids pH Electrodes > InLab Solids Go-ISM Electrode

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.