missing translation for 'onlineSavingsMsg'
Learn More

Oxoid One Broth - Salmonella 10 Pk

One Broth - Salmonella 10 Pk

Brand:  Oxoid BO1096S

75.53 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 13215479

  • £106.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

One Broth - Salmonella 10 Pk

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4500564 Certificate TypeCertificate of Analysis Date01/04/2025 Catalog NumberBO1096S
Lot Number4500562 Certificate TypeCertificate of Analysis Date01/04/2025 Catalog NumberBO1096S
Lot Number4500558 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog NumberBO1096S
Lot Number4500560 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog NumberBO1096S
Lot Number4497099 Certificate TypeCertificate of Analysis Date18/03/2025 Catalog NumberBO1096S
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oxoid One Broth - Salmonella 10 Pk >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.