Learn More
Thermo Scientific™ ONE Broth-Salmonella Base
Enrich and recover Salmonella spp. in a single step with Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Base.
Brand: Thermo Scientific™ CM1091B
831.40 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Enrich and recover Salmonella spp. from food samples in a single step with Thermo Scientific™ Oxoid™ ONE Broth-Salmonella Base, when prepared with Thermo Scientific Oxoid ONE Broth-Salmonella Selective Supplement (SR0242).
ONE Broth-Salmonella Base is a highly nutritious and selective medium that allows recovery of Salmonella species within 16 to 20 hours. ONE Broth-Salmonella Base requires only a single enrichment medium step. When followed by inoculation onto Thermo Scientific™ Brilliance™ Salmonella Agar and the Thermo Scientific Oxoid Salmonella Latex Test, it forms the Oxoid™ Salmonella Precis™ method which reduces time to result over conventional culture methods. The Oxoid Salmonella Precis method is AFNOR validated.
Specifications
10°C to 30°C | |
Powder | |
500 g |
ONE Broth-Salmonella Base | |
Dehydrated Culture Media |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6246556 | Certificate of Analysis | 25/03/2025 | CM1091B |
6232886 | Certificate of Analysis | 25/02/2025 | CM1091B |
6183194 | Certificate of Analysis | 06/11/2024 | CM1091B |
6168756 | Certificate of Analysis | 18/10/2024 | CM1091B |
6148227 | Certificate of Analysis | 02/08/2024 | CM1091B |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.