missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Surface pH Electrodes
A pH electrode featuring a flat, rugged plastic membrane for measuring samples with flat surfaces.
£327.00 - £488.00
Specifications
Junction | Ceramic |
---|---|
pH Range | 1 to 11 |
Membrane | LoT Glass |
Temperature (Metric) Range | 0°C to 50°C |
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | |||
10049102
|
METTLER TOLEDO™
51343157 |
InLab™ Surface | Combination Electrode | S7 | S7 to BNC (30281915) |
£327.00
Each |
In Stock
Log in to see stock. ![]() |
|||
15539611
|
METTLER TOLEDO™
30249570 |
InLab™ Surface Pro-ISM | Combination Electrode; Integrated Temperature Probe; Intelligent Sensor Management (ISM) Technology | Multi-Pin | Multi-Pin to BNC/RCA (30281896) |
£488.00
Each |
In Stock
Log in to see stock. |
|||
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Agar plates
- Drop size samples
- Paper
- Skin
- Leather
- Suspended solids
- Textiles and Prints
Accessories & Care
- Ceramic ring junction
- Polysulfone membrane
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Ceramic | |
1 to 11 | |
LoT Glass | |
0°C to 50°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
METTLER TOLEDO™ InLab™ Surface pH Electrodes