missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Innova™ 40R Incubator Shaker

Use these versatile incubator shakers for support high-speed applications and heavy workloads. New Brunswick Scientific™ Innova™ 40R Incubator Shakers provide very broad temperature capabilities for culturing a wide variety of organisms in flasks up to 3 liters.

Brand:  Eppendorf™ M1299-0086

13381.81 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Product Code. 12518886

  • £14086.12 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Exceptionally quiet operation provides a favorable work environment
  • Triple-eccentric counterbalanced drive in cast iron housing provides vibration and trouble free operation for years
  • Compact benchtop model – fits easily on the bench and holds flasks up to 3L
  • Clear cover allows easy viewing of the cultures and lifts easily to access all samples
  • Versatile accessory platform (sold separately) accommodates flasks up to 3L as well as test tube racks and Microplate holders
  • Certified. Meets cETL, UL, CSA and CE standards
TRUSTED_SUSTAINABILITY
Specifications

Specifications

19 mm
230 V, 50 Hz
3 L
50 Hz
101.6 cm
Innova 40R
25 to 500 rpm
230 V
Incubator Shaker
76.2 cm
55.9 cm
0.01 to 99:59 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers