Learn More
Gosselin™ Inoculating T-Shaped Spreaders
For spreading over the surface of solid media in microbiological applications. Gosselin™ Inoculating T-Shaped Spreaders are manufactured from polystyrene plastic.
Brand: Gosselin™ ETAT-03
Code : A0
Additional Details : Weight : 1.00000kg
Description
- More precise
- Sterile and disposable
- Packed in tamper-evident ZIP bags
Specifications
Yes | |
T-Shape | |
Pack of 1500 |
Sterile | |
Green | |
148 mm |