missing translation for 'onlineSavingsMsg'
Learn More

Inosine 5'-monophophate disodium salt, ≥98%, MP Biomedicals™

£33.10

Chemical Identifiers

CAS 4691-65-0
Molecular Formula C10H11N4Na2O8P
Molecular Weight (g/mol) 392.17
MDL Number MFCD00036201,MFCD00036201
InChI Key AANLCWYVVNBGEE-IDIVVRGQSA-L
Synonym disodium 5'-inosinate
PubChem CID 123134061
IUPAC Name disodium [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1NC=NC2=O
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11328889
View Documents
MP Biomedicals
0210205005
5 g
£33.10
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

4691-65-0
392.17
AANLCWYVVNBGEE-IDIVVRGQSA-L
123134061
[Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1NC=NC2=O
C10H11N4Na2O8P
MFCD00036201,MFCD00036201
disodium 5'-inosinate
disodium [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methyl phosphate

Specifications

4691-65-0
C10H11N4Na2O8P
disodium 5'-inosinate
[Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1NC=NC2=O
392.17
392.2
Inosine 5'-monophophate disodium salt
White
MFCD00036201,MFCD00036201
AANLCWYVVNBGEE-IDIVVRGQSA-L
disodium [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methyl phosphate
123134061
Crystalline Powder
Videos
SDS
Documents

Documents

Product Certifications