missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ V4A Stainless Steel Inserts for Novartos Multi

The V4A Stainless Steel Inserts for Buerkle™ Novartos Multi fit precisely and can easily be replaced. Inserts are available in volumes from 0 to 2.0 ml.

Brand:  Buerkle™ 5358-0101

178.81 GBP valid until 2024-12-31
Use promo code "23267" to get your promotional price.



Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15993247

  • £193.00 / Each
Estimated Shipment: 29-07-2024
to see stock.
Add to basket

Description

Description

  • Fit precisely
  • Can easily be replaced
  • Stainless steel AISI 316L
Specifications

Specifications

Insert
Autoclavable
FDA
Stainless Steel (V4A)
0.1 mL
Novartos Multi Sampler
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions