missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ INSTA-Blot Multi-species Liver, Lung, Spleen Tissues
PVDF membrane containing denatured protein from cells or tissue lysates
Brand: Novus Biologicals™ NBP2-30117
Additional Details : Weight : 0.00970kg
Description
- Screening proteins from various human, mouse and rat tissues
- Contains 11 human, mouse and rat tissue lysates (20μg total protein per lane) on 4-20% Tris-Glycine mini gel
- Prepared from resolution by SDS-PAGE at 125V for 1 hour
- Proteins are electro-blotted for 2 hours at 25V onto PVDF membrane
- Stained with amido black for visualization of proteins
Recommended Storage
Store at room temperature
Specifications
Screening proteins from various human, mouse and rat tissues | |
1 Membrane | |
PVDF |