missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Installation Kits for Packed and Micropacked Columns

Installation kits for packed and micropacked columns for direct injections or valve applications.

Brand:  Restek™ 21067

Encompass_Preferred

 View more versions of this product

Product Code. 13194462

  • £61.71 / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket

Includes:

Part No. 21065

One 1/16 in. Valco nut, one 1/16 in. stainless steel nut, one 1/16 in. Vespel/graphite ferrule, one 1/16 in. graphite ferrule, one stainless steel ferrule, one 1/16 in. stainless steel front ferrule, one 1/16 in. stainless steel back ferrule

Part No. 21066

Two 1/16 in. stainless steel nuts, two 1/16 in. Vespel/graphite ferrules, two 1/16 in. graphite ferrules, two 1/16 in. stainless steel front ferrules, two 1/16 in. stainless steel back ferrules

Part No. 21067

One 1/8 in. stainless steel nut, one stainless steel Valco nut, one 1/8 in. Vespel/graphite ferrule, one stainless steel Valco ferrule, one 1/8 in. stainless steel front ferrule, one 1/8 in. stainless steel back ferrule

This item is not returnable. View return policy

Description

Description

Part No. 21065
  • For micropacked columns
  • For 1mm ID columns
  • For valve applications
Part No. 21066
  • For micropacked columns
  • For 1mm ID columns
  • For direct injections
Part No. 21067
  • For packed columns
  • For 2mm ID columns
  • For valve applications
Specifications

Specifications

Installation Kit
For Valve Applications
2 mm I.D. Columns
2 mm
One 1/8 in. stainless steel nut, one stainless steel Valco nut, one 1/8 in. Vespel/graphite ferrule, one stainless steel Valco ferrule, one 1/8 in. stainless steel front ferrule, one 1/8 in. stainless steel back ferrule
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.