missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Installation Kits for Packed and Micropacked Columns

Installation kits for packed and micropacked columns for direct injections or valve applications.

£61.71 - £95.59

Specifications

Type Installation Kit
View More Specs

Products 3
Product Code Brand Diameter (Metric) Inner For Use With (Application) For Use With Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner For Use With (Application) For Use With Price Quantity & Availability  
13184462
Encompass_Preferred
View Documents
Restek™
21066
1 mm For Direct Injections 1 mm O.D. Columns
£95.59
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
13194462
Encompass_Preferred
View Documents
Restek™
21067
2 mm For Valve Applications 2 mm I.D. Columns
£61.71
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
13174462
Encompass_Preferred
View Documents
Restek™
21065
1 mm For Valve Applications 1 mm O.D. Columns
£73.81
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
Description

Description

Part No. 21065
  • For micropacked columns
  • For 1mm ID columns
  • For valve applications
Part No. 21066
  • For micropacked columns
  • For 1mm ID columns
  • For direct injections
Part No. 21067
  • For packed columns
  • For 2mm ID columns
  • For valve applications
Specifications

Specifications

Installation Kit
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.