missing translation for 'onlineSavingsMsg'
Learn More

3M™ Instant Microbe Removing Wipes

Trap, hold and remove microbes with these cleaning wipes to virtually eliminate cross contamination. They work using just water, removing microbes instantly as you wipe. Great for high-touch, fast-turnaround areas.

Brand:  3M™ 7100244448

Additional Details : Weight : 1.10800kg

 View more versions of this product

Product Code. 17751679

  • £93.05 / 4 canisters
Estimated Shipment: 15-08-2024
to see stock.
Add to basket

Description

Description

  • Quickly traps, holds and removes 99.9% of microbes, including SARS-CoV-2*
  • Virtually eliminates cross contamination
  • Works using only water – no disinfectant or specific PPE required
  • Easily removes as you wipe, no dwell time
  • Also works with quat disinfectants
  • Refillable containers to reduce waste
  • If wipes dry out, simply add water
Specifications

Specifications

Wipe
195 mm
254 mm
195 mm x 254 mm
85 Sheets/Canister, 4 Canisters/Case
7.7 in.
10 in.
EN/SP, OUS
Microbe Removal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

*Based on third party testing with water only.