Learn More
Terumo™ Insulin Syringe
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ Insulin Syringes feature a unique double beveled design for greater patient comfort.
Brand: Terumo™ 8MDSS01S2613E
This item is not returnable.
View return policy
Description
- Ultra-sharp needles
- Clear barrels
- Bold, precise scale markings
- Smooth plunger action
- Tamper evident peel packs
- Sterile, Individually wrapped
Specifications
26 G | |
Terumo Insulin Syringe | |
1 mL |
13 mm | |
100 Pack | |
Sterile, Individually Wrapped |
Product Content Correction
Product Title
Terumo™ Insulin Syringe > Volume: 1 mL
Spot an opportunity for improvement?Share a Content Correction