missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin beta 6 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
£153.34 - £437.13
Specifications
Antigen | Integrin beta 6 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18798167
|
R&D Systems
AF2389-SP |
25 μg |
£153.34
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18092282
|
R&D Systems
AF2389 |
100 μg |
£437.13
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin beta 6 Polyclonal specifically detects Integrin beta 6 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Integrin beta 6 | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
integrin beta-6, integrin, beta 6, ITGB6 | |
ITGB6 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Q9Z0T9 | |
3694 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 6 Gly22-Asn706 Accession # Q9Z0T9 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Integrin beta 6 in direct ELISAs and Western blots. |
For Research Use Only