missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Interior IP68 Socket for UF30
Interior IP68 socket that can be turned on and off with a switch.
Brand: Memmert™ UF30R3
This item is not returnable.
View return policy
Description
- Can only be ordered with limited temperature range - max. +60°C)
- Electrical Requirements: 230 V, 2 A, 500 W
- Can be switched off with the On/Off switch
- Cannot be switched individually
- Moisture tight IP68
- Requires option A8 against surcharge
Specifications
UF30 Oven | |
Interior Socket | |
230 V | |
2 A |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Interior IP68 Socket for UF30 > For Use With: UF30
Spot an opportunity for improvement?Share a Content Correction